Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPA2, p Ser33 Antibody, Novus Biologicals™
SDP

Catalog No. NB100544 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100544 Supplier Novus Biologicals Supplier No. NB100544
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 17 publications

RPA2 (p Ser33) Polyclonal specifically detects RPA2 (p Ser33) in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Simple Western, Western Blot.

Specifications

Antigen RPA2 (p Ser33)
Applications Immunocytochemistry, Immunofluorescence, Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:5000, Simple Western, Immunocytochemistry/Immunofluorescence 1:500 - 1:2500
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. P15927
Gene Alias REPA2, Replication factor A protein 2, replication protein A 32 kDa subunit, Replication protein A 34 kDa subunit, replication protein A2 (32kD), replication protein A2, 32kDa, RF-A protein 2, RP-A p32, RP-A p34, RPA32, RPA34
Gene Symbols RPA2
Host Species Rabbit
Immunogen Immunogen was a phosphorylated synthetic peptide, which represented a portion of human replication protein A2, 32 kDa surrounding phosphorylated serine that corresponded to position 33 using the numbering given in entry NP_002937.1 (GeneID 6118).
Molecular Weight of Antigen 32 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Nucleotide Excision Repair, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6118
Test Specificity NB100-544 is specific for human RPA-32 pSer33 protein.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.