Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPA70 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220206 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220206 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220206 Supplier Novus Biologicals Supplier No. NBP220206

Rabbit Polyclonal Antibody

RPA70 Polyclonal specifically detects RPA70 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RPA70
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.43mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. P27694
Gene Alias HSSB, MST075, MSTP075, REPA1RF-A protein 1, Replication factor A protein 1, replication protein A 70 kDa DNA-binding subunit, replication protein A1 (70kD), replication protein A1, 70kDa, RF-A, RP-A, RPA70RP-A p70, Single-stranded DNA-binding protein
Gene Symbols RPA1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RPA70. The exact sequence is proprietary.
Molecular Weight of Antigen 68 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Nucleotide Excision Repair
Primary or Secondary Primary
Gene ID (Entrez) 6117
Test Specificity Human. Species reactivity based on sequence identity: Mouse (89%), Bovine (93%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.