Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPC62 Antibody (OTI2H1), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP273942UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273942UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273942UV Supplier Novus Biologicals Supplier No. NBP273942UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RPC62 Monoclonal specifically detects RPC62 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RPC62
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2H1
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DNA-directed III 62 kDa polypeptide, DNA-directed RNA polymerase III subunit C, DNA-directed RNA polymerase III subunit RPC3, polymerase (RNA) III (DNA directed) polypeptide C (62kD), RNA polymerase III 62 kDa subunit, RNA polymerase III subunit C3, RPC62RPC3
Gene Symbols POLR3C
Host Species Mouse
Immunogen Full length recombinant protein of human RPC62 (NP_006459) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10623.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.