Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPE65 Antibody (401.8B11.3D9), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB100355G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100355G 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100355G Supplier Novus Biologicals Supplier No. NB100355G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RPE65 Monoclonal specifically detects RPE65 in Bovine, Canine, Chicken, Human, Mouse, Pig, Primate, Rat, Xenopus species samples. It is validated for CyTOF, Flow Cytometry, Gene Knock-Out, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vivo Assay, Western Blot.

Specifications

Antigen RPE65
Applications CyTOF, Flow Cytometry, Gene Knock-Out, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In vivo Assay, Western Blot
Classification Monoclonal
Clone 401.8B11.3D9
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias All-trans-retinyl-palmitate hydrolase, EC 3.1.1.64, EC 5.2.1.7, LCA2p63, mRPE65, RBP-binding membrane protein, rd12, Retinal pigment epithelium-specific 65 kDa protein, retinal pigment epithelium-specific protein (65kD), retinal pigment epithelium-specific protein 65kDa, retinitis pigmentosa 20 (autosomal recessive), retinoid isomerohydrolase, Retinol isomerase, RP20, sRPE65
Gene Symbols RPE65
Host Species Mouse
Immunogen This RPE65 Antibody (401.8B11.3D9) was developed against bovine RPE65 microsomal membrane proteins. [UniProt# Q28175]
Molecular Weight of Antigen 65 kDa
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Sensory Systems, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 6121
Test Specificity NB100-355 is specific for RPE65.
Target Species Bovine, Canine, Chicken, Human, Mouse, Pig, Primate, Rat, Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.