Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPGRIP1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | RPGRIP1 |
|---|---|
| Concentration | 0.41 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8578
![]()
|
Proteintech Group Inc
132141AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a photoreceptor protein that interacts with retinitis pigmentosa GTPase regulator protein and is a key component of cone and rod photoreceptor cells. Mutations in this gene lead to autosomal recessive congenital blindness.Specifications
| RPGRIP1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q96KN7 | |
| 57096 | |
| RPGRIP1 Fusion Protein Ag4089 | |
| Primary | |
| -20°C | |
| RPGRIP1 |
| 0.41 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CORD9, LCA6, RGI1, RGRIP, RPGR interacting protein 1, RPGRIP, RPGRIP1, RPGRIP1d | |
| RPGRIP1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title