Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPIP8 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB043683 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB043683 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB043683 Supplier Novus Biologicals Supplier No. NBP297453UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPIP8 Polyclonal specifically detects RPIP8 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RPIP8
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 350
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Rap2-interacting protein 8, RAP2IPRUN domain-containing protein 3A, RPIP-8, RPIP8RaP2 interacting protein 8, RUN domain containing 3A
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RPIP8 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10900
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.