Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL10 Antibody (OTI6B11), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP273944A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273944A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273944A70 Supplier Novus Biologicals Supplier No. NBP273944AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RPL10 Monoclonal specifically detects RPL10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RPL10
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6B11
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 60S ribosomal protein L10, DXS648EFLJ27072, DXS648FLJ23544, Laminin receptor homolog, NOV, Protein QM, QMDKFZp686J1851, ribosomal protein L10, Tumor suppressor QM, Wilms tumor-related protein
Gene Symbols RPL10
Host Species Mouse
Immunogen Full length recombinant protein of human RPL10 (NP_006004) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6134
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.