Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL11 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB290322 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290322 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290322 Supplier Novus Biologicals Supplier No. NBP222283APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPL11 Polyclonal antibody specifically detects RPL11 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative)

Specifications

Antigen RPL11
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation (Negative)
Formulation PBS
Gene Alias cell growth-inhibiting protein 34,60S ribosomal protein L11, CLL-associated antigen KW-12, DBA7, GIG34, ribosomal protein L11
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 128 and 178 of human Ribosomal Protein L11 using the numbering given in entry NP_000966.2 (GeneID 6135).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6135
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.