Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL13A Antibody, Novus Biologicals™
SDP

Catalog No. NBP155222 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP155222 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP155222 Supplier Novus Biologicals Supplier No. NBP155222
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPL13A Polyclonal specifically detects RPL13A in Human samples. It is validated for Western Blot.

Specifications

Antigen RPL13A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P40429
Gene Alias 60S ribosomal protein L13a, ribosomal protein L13a, tissue specific transplantation antigen 1,23 kDa highly basic protein, TSTA1
Gene Symbols RPL13A
Host Species Rabbit
Immunogen Synthetic peptides corresponding to RPL13A(ribosomal protein L13a) The peptide sequence was selected from the middle region of RPL13A. Peptide sequence HEVGWKYQAVTATLEEKRKEKAKIHYRKKKQLMRLRKQAEKNVEKKIDKY.
Molecular Weight of Antigen 22 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23521
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.