Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL18A Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB330031 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB330031 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB330031 Supplier Novus Biologicals Supplier No. NBP297781JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPL18A Polyclonal antibody specifically detects RPL18A in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen RPL18A
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias 60S ribosomal protein L18a, ribosomal protein L18a, ribosomal protein L18a-like protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RPL18A fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6142
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.