Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL22 Antibody, Novus Biologicals™
SDP

Catalog No. NBP106069 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106069 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106069 Supplier Novus Biologicals Supplier No. NBP106069
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 6 publications

RPL22 Polyclonal specifically detects RPL22 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen RPL22
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 3 - 6 ug/mL, Immunohistochemistry-Paraffin 3-6 ug/ml, Peptide ELISA detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P35268
Gene Alias EAPHBP15, EBER-associated protein, Epstein-Barr virus small RNA-associated protein, Epstein-Barr-encoded RNA-associated protein, HBP15/L22, heparin-binding protein 15, Heparin-binding protein HBp15, ribosomal protein L22,60S ribosomal protein L22
Gene Symbols RPL22
Host Species Goat
Immunogen Peptide with sequence C-SKESYELRYFQINQ corresponding to C-Terminus according to NP_000974.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6146
Test Specificity Human. Predicted cross-reactivity based on sequence identity: Mouse, Rat.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.