Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL24/RLP24 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15547920 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15547920 20 μL
NBP155479 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15547920 Supplier Novus Biologicals Supplier No. NBP15547920UL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

RPL24/RLP24 Polyclonal specifically detects RPL24/RLP24 in Human samples. It is validated for Western Blot.

Specifications

Antigen RPL24/RLP24
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9UHA3
Gene Alias C15orf15, HRP-L30-iso, L30, RLP24, RPL24, RPL24L, RSL24D1 ribosomal L24 domain containing 1, TVAS3
Gene Symbols RSL24D1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to C15ORF15 The peptide sequence was selected from the middle region of C15ORF15. Peptide sequence FIMNRLKKNKELQKVQDIKEVKQNIHLIRAPLAGKGKQLEEKMVQQLQED.
Molecular Weight of Antigen 19 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51187
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.