Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL36 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB330860 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB330860 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB330860 Supplier Novus Biologicals Supplier No. NBP297618MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPL36 Polyclonal antibody specifically detects RPL36 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen RPL36
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DKFZp566B023,60S ribosomal protein L36, M, ribosomal protein L36
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RPL36 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25873
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.