Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPL36A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $646.00
Specifications
Antigen | RPL36A |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
RPL36A Polyclonal specifically detects RPL36A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RPL36A | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
6173 | |
This antibody was developed against a recombinant protein corresponding to amino acids: MIAPTDSHEEVRSGTSYILPFASRFLSFRA | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
60S ribosomal protein L36a, Cell growth-inhibiting gene 15 protein, Cell migration-inducing gene 6 protein, dJ164F3.3 (ribosomal protein L44), L44L, L44-like ribosomal protein, MGC72020, MIG6,60S ribosomal protein L44, ribosomal protein L36a, ribosomal protein L44, RPL36AL, RPL44 | |
RPL36A | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title