Learn More
Description
Specifications
Specifications
| Antigen | RPL37A |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 60S ribosomal protein L37a, MGC74786, ribosomal protein L37a |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human RPL37A fragment. |
| Purification Method | Antigen Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
