Learn More
Invitrogen™ RPL37A Polyclonal Antibody

Description
Sequence homology: Cow: 100%; Dog: 100%; Goat: 79%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Specifications
Specifications
| Antigen | RPL37A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | RPL37A |
| Gene Accession No. | P61513 |
| Gene Alias | 60S ribosomal protein L37a; L37A; Large ribosomal subunit protein eL43; ribosomal protein L37a; ribosomal protein L37a-like; Rpl37a |
| Gene Symbols | RPL37A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.