Learn More
Invitrogen™ RPL6 Polyclonal Antibody

Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 86%; Human: 100%; Mouse: 87%; Rat: 85%.
Specifications
Specifications
| Antigen | RPL6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | RPL6 |
| Gene Accession No. | Q02878 |
| Gene Alias | 60S ribosomal protein L6; DNA-binding protein TAXREB107; L6; Large ribosomal subunit protein eL6; Neoplasm-related protein C140; ribosomal protein L6; Rpl6; SHUJUN-2; Taxreb107; TAX-responsive enhancer element-binding protein 107; TXREB1 |
| Gene Symbols | RPL6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.