Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL7 Antibody, Novus Biologicals™
SDP

Catalog No. NB1002268 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002268 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002268 Supplier Novus Biologicals Supplier No. NB1002268
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

RPL7 Polyclonal specifically detects RPL7 in Human, Mouse samples. It is validated for Immunoprecipitation (Negative), Western Blot.

Specifications

Antigen RPL7
Applications Immunoprecipitation (Negative), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. P18124
Gene Alias humL7-1,60S ribosomal protein L7, MGC117326, ribosomal protein L7
Gene Symbols RPL7
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human Ribosomal Protein 7 using the numbering given in entry NP_000962.2 (GeneID 6129).
Molecular Weight of Antigen 29 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6129
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with X. laevis, X. tropicalis, Panda, Orangutan, Rhesus Monkey, Gorilla, Sea lamprey, West Indian ocean coelacanth and Spotted green pufferfish.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.