Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPL7 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB292452 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292452 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292452 Supplier Novus Biologicals Supplier No. NB1002268PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPL7 Polyclonal antibody specifically detects RPL7 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative)

Specifications

Antigen RPL7
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation (Negative)
Formulation PBS
Gene Alias humL7-1,60S ribosomal protein L7, MGC117326, ribosomal protein L7
Host Species Rabbit
Immunogen Maps to a region between residue 75 and 125 of human Ribosomal Protein 7 using the numbering given in entry NP_000962.2 (GeneID 6129).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6129
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.