Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPLP0 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB047594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB047594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB047594 Supplier Novus Biologicals Supplier No. NBP297776AF700

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

RPLP0 Polyclonal specifically detects RPLP0 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RPLP0
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 60S acidic ribosomal protein P0, 60S ribosomal protein L10E, acidic ribosomal phosphoprotein P0, L10E, MGC111226, MGC88175, P0, PRLP0, ribosomal protein, large, P0, RPP0
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RPLP0 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6175
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.