Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

rpmB/L28 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB287495 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287495 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287495 Supplier Novus Biologicals Supplier No. NBP323514AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

rpmB/L28 Polyclonal antibody specifically detects rpmB/L28 in E. coli samples. It is validated for Western Blot, ELISA

Specifications

Antigen rpmB/L28
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias 50S ribosomal subunit protein L28, ECK3627, Large ribosomal subunit protein bL28, rpmB
Host Species Rabbit
Immunogen Antibody was raised against an 15 amino acid peptide near the amino terminus of E. coli rpmB/L28. The immunogen is located within the first 50 amino acids of rpmB/L28.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 946941
Target Species E. coli
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.