Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

rpmH/L34 Antibody - C-terminus - BSA Free, Novus Biologicals™
SDP

Catalog No. NB287541 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287541 0.1 mg
NB287543 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB287541 Supplier Novus Biologicals Supplier No. NBP3235160.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

rpmH/L34 Polyclonal antibody specifically detects rpmH/L34 in E. coli samples. It is validated for Western Blot, ELISA

Specifications

Antigen rpmH/L34
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml, ELISA
Formulation PBS
Gene Alias 50S ribosomal subunit protein L34, ECK3695, Large ribosomal subunit protein bL34, rimA, rpmH, ssaF
Host Species Rabbit
Immunogen Antibody was raised against an 15 amino acid peptide near the carboxyl terminus of E. coli rpmH/L34. The immunogen is located within the last 30 amino acids of rpmH/L34.
Purification Method Affinity purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 948216
Target Species E. coli
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.