Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPP38 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 128741AP150UL

Description
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5′-ends. RPP38 may associate transiently with RNase P RNA as a factor involved in the transport of H1 RNA to the putative site of its assembly in the cell, the nucleolus.Specifications
RPP38 | |
Polyclonal | |
Unconjugated | |
RPP38 | |
RNaseP protein p38, RP11 455B2.5, RPP38 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10557, 227522, 291317 | |
-20°C | |
Liquid |
Western Blot | |
0.61 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P78345, Q80UU2 | |
Rpp38 | |
RPP38 Fusion Protein Ag3552 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction