Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPP38 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | RPP38 |
---|---|
Concentration | 0.61 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8318
![]() ![]() |
Proteintech Group Inc
128741AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Component of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5′-ends. RPP38 may associate transiently with RNase P RNA as a factor involved in the transport of H1 RNA to the putative site of its assembly in the cell, the nucleolus.Specifications
RPP38 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P78345, Q80UU2 | |
10557, 227522, 291317 | |
RPP38 Fusion Protein Ag3552 | |
Primary | |
-20°C | |
RPP38 |
0.61 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
RNaseP protein p38, RP11 455B2.5, RPP38 | |
Rpp38 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title