Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RPRD1B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321336100UL
Description
RPRD1B Polyclonal antibody specifically detects RPRD1B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
RPRD1B | |
Polyclonal | |
Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Cell cycle-related and expression-elevated protein in tumor, cell-cycle related and expression-elevated protein in tumor, chromosome 20 open reading frame 77, CREPTC20orf77, dJ1057B20.2, DKFZp434P0735, FLJ44520, NET60, regulation of nuclear pre-mRNA domain containing 1B, regulation of nuclear pre-mRNA domain-containing protein 1B | |
This antibody was developed against Recombinant Protein corresponding to amino acids: RLAAELEDRRQLARMLVEYTQNQKDVLSEKEKK | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
58490 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction