Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPRD2 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB290743 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290743 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290743 Supplier Novus Biologicals Supplier No. NBP197932APC
Add to Cart
Add to Cart

Goat Polyclonal Antibody

RPRD2 Polyclonal antibody specifically detects RPRD2 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen RPRD2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias HSPC099, KIAA0460FLJ32145, regulation of nuclear pre-mRNA domain containing 2, regulation of nuclear pre-mRNA domain-containing protein 2
Host Species Goat
Immunogen The immunogen this antibody was made to, maps to a region between residues 1000 and 1050 of human RPRD2 using the numbering given in TrEMBL entry Q5VT52 (GeneID 23248).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 23248
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.