Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS2 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB290472 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290472 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290472 Supplier Novus Biologicals Supplier No. NBP214852APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPS2 Polyclonal antibody specifically detects RPS2 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen RPS2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias LLREP3, MGC102851, MGC117344, MGC117345,40S ribosomal protein S4, OK/KNS-cl.6,40S ribosomal protein S2, Protein LLRep3, ribosomal protein S2, RPS4
Host Species Rabbit
Immunogen This antibody maps to a region between residue 50 and 100 of human Ribosomal Protein S2 using the numbering given in entry NP_002943.2 (GeneID 6187).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6187
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.