Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS23 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB330842 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB330842 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB330842 Supplier Novus Biologicals Supplier No. NBP297621PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPS23 Polyclonal antibody specifically detects RPS23 in Human samples. It is validated for Western Blot

Specifications

Antigen RPS23
Applications Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias FLJ35016, homolog of yeast ribosomal protein S28, ribosomal protein S23,40S ribosomal protein S23
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RPS23 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6228
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.