Learn More
Description
Specifically detects RPS25 in Human samples, and it is validated for Western Blotting
Specifications
Specifications
| Antigen | RPS25 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000 |
| Formulation | 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal |
| Gene Alias | ribosomal protein S25,40S ribosomal protein S25 |
| Gene Symbols | RPS25 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of human RPS25. The exact sequence is proprietary. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
