Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS4X Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB362908 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362908 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362908 Supplier Novus Biologicals Supplier No. NBP321449AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RPS4X Polyclonal antibody specifically detects RPS4X in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen RPS4X
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CCG2Single copy abundant mRNA protein, DXS306, FLJ40595, ribosomal protein S4, X-linked, RPS4cell cycle gene 2, SCAR40S ribosomal protein S4, X isoform, SCR10ribosomal protein S4X isoform, single-copy abundant mRNA
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RPS4X fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6191
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.