Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPS6KC1 Antibody (OTI10F11), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP273946R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273946R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273946R Supplier Novus Biologicals Supplier No. NBP273946R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RPS6KC1 Monoclonal specifically detects RPS6KC1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RPS6KC1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI10F11
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.11.1, humS6PKh1, ribosomal protein S6 kinase delta-1, ribosomal protein S6 kinase, 52kD, polypeptide 1,52 kDa ribosomal protein S6 kinase, ribosomal protein S6 kinase, 52kDa, polypeptide 1, Ribosomal S6 kinase-like protein with two PSK domains 118 kDa protein, RPK118, S6K-delta-1, SPHK1-binding protein
Gene Symbols RPS6KC1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-305 of human RPS6KC1 (NP_036556) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 26750.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.