Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RPTOR Antibody (514208), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB377255 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377255 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377255 Supplier Novus Biologicals Supplier No. FAB5957I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RPTOR Monoclonal specifically detects RPTOR in Human, Mouse samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen RPTOR
Applications CyTOF, Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 514208
Conjugate Janelia Fluor 549
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias KIAA1303p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats, KOG1, Mip1, p150 target of rapamycin (TOR)-scaffold protein, Raptor, regulatory associated protein of MTOR, complex 1, regulatory-associated protein of mTOR
Host Species Mouse
Immunogen E. coli-derived recombinant human RPTOR, Lys77-Gln230, Accession # Q8N122
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Hypoxia, mTOR Pathway, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57521
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.