Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRAD Antibody, Novus Biologicals™
SDP

Catalog No. NBP227500 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227500 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227500 Supplier Novus Biologicals Supplier No. NBP227500
Add to Cart
Add to Cart

Goat Polyclonal Antibody

RRAD Polyclonal specifically detects RRAD in Mouse samples. It is validated for Western Blot, Peptide ELISA, Knockout Validated.

Specifications

Antigen RRAD
Applications Western Blot, ELISA, Immunoassay
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 2 ug/ml, Peptide ELISA Detection limit 1:64000, Knockout Validated
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_004156.1
Gene Alias RAD1, RADGTP-binding protein RAD, RAS (RAD and GEM) like GTP binding 3, Ras associated with diabetes, Ras-related associated with diabetes, REM3
Gene Symbols RRAD
Host Species Goat
Immunogen Peptide with sequence C-HRRSMPVDERDLQ corresponding to internal region according to NP_004156.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6236
Test Specificity Reported variants represent identical protein: NP_004156.1, NP_001122322.1
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.