Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRAGC Rabbit anti-Human, DyLight 405, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB046044 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB046044 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB046044 Supplier Novus Biologicals Supplier No. NBP297651V

Rabbit Polyclonal Antibody

RRAGC Polyclonal specifically detects RRAGC in Human samples. It is validated for Western Blot.

Specifications

Antigen RRAGC
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ13311, GTPase-interacting protein 2, GTR2, Rag C, Rag C protein, RAGC, Ras-related GTP binding C, ras-related GTP-binding protein C, TIB929
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RRAGC fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 64121
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.