Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRAGD Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB290040 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290040 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290040 Supplier Novus Biologicals Supplier No. NBP232106APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RRAGD Polyclonal antibody specifically detects RRAGD in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen RRAGD
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias bA11D8.2.1, DKFZP761H171, FLJ44176, Rag D, Rag D protein, RAGD, Ras-related GTP binding D, ras-related GTP-binding protein D
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Ras-Related GTP Binding D using the numbering given in entry NP_067067.1 (GeneID 58528).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 58528
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.