Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RRAGD Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156669
Description
RRAGD Polyclonal specifically detects RRAGD in Human samples. It is validated for Western Blot.Specifications
RRAGD | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
bA11D8.2.1, DKFZP761H171, FLJ44176, Rag D, Rag D protein, RAGD, Ras-related GTP binding D, ras-related GTP-binding protein D | |
Rabbit | |
45 kDa | |
100 μL | |
Cancer, mTOR Pathway | |
58528 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9NQL2 | |
RRAGD | |
Synthetic peptides corresponding to RRAGD (Ras-related GTP binding D) The peptide sequence was selected from the middle region of RRAGD)(50ug). Peptide sequence CDMIDVVIDISCIYGLKEDGAGTPYDKESTAIIKLNNTTVLYLKEVTKFL. | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Sheep: 77%; Zebrafish: 77%; Guinea pig: 77%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Sheep, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction