Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRM1 Antibody (OTI5G5), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP273947A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273947A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273947A32 Supplier Novus Biologicals Supplier No. NBP273947AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RRM1 Monoclonal specifically detects RRM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RRM1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5G5
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 1.17.4.1, R1, ribonucleoside-diphosphate reductase large subunit, Ribonucleoside-diphosphate reductase subunit M1, Ribonucleotide reductase large subunit, ribonucleotide reductase M1, ribonucleotide reductase M1 polypeptide, ribonucleotide reductase, large subunit, ribonucleotide reductase, R1 subunit, RIR1, RR1
Gene Symbols RRM1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 541-792 of human RRM1 (NP_001024) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6240
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.