Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RRM1 Antibody (RRM1/4372R), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB381482 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381482 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381482 Supplier Novus Biologicals Supplier No. NBP308331AF350
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

RRM1 Monoclonal antibody specifically detects RRM1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen RRM1
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone RRM1/4372R
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias EC 1.17.4.1, R1, ribonucleoside-diphosphate reductase large subunit, Ribonucleoside-diphosphate reductase subunit M1, Ribonucleotide reductase large subunit, ribonucleotide reductase M1, ribonucleotide reductase M1 polypeptide, ribonucleotide reductase, large subunit, ribonucleotide reductase, R1 subunit, RIR1, RR1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues within aa700-800 of human RRM1 (exact sequence is proprietary) (Uniprot: IDP23921)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6240
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.