Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

rRNA Antibody (Y10b), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB10662A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10662A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10662A750 Supplier Novus Biologicals Supplier No. NB100662AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

rRNA Monoclonal specifically detects rRNA in All species, Bacteria, Human, Mouse samples. It is validated for CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation.

Specifications

Antigen rRNA
Applications CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone Y10b
Conjugate Alexa Fluor 750
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular)
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ribosomal RNA 5.8s
Host Species Mouse
Immunogen The whole 5.8S ribosomal RNA.
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity This is specific for ribosomal RNA. There is no cross-reactivity with other RNAs.
Target Species All species, Bacteria, Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.