Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

rRNA Antibody (Y10b) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB100662SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100662SS 0.025 mL
NB100662 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100662SS Supplier Novus Biologicals Supplier No. NB100662SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 21 publication

rRNA Monoclonal specifically detects rRNA in All species, Bacteria, Chicken, Fish, Human, Mouse, Plant, Rat samples. It is validated for ChIP Assay, CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen rRNA
Applications ChIP Assay, CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Y10b
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/ml, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34980897), Immunoprecipitation 10 ul/lysate, Immunohistochemistry-Paraffin 1:100 - 1:300, Flow (Intracellular) 2.5 ug/ml, Immunocytochemistry 1 - 3 ug/ml, CyTOF-reported, CyTOF-ready reported by customer review
Gene Alias ribosomal RNA 5.8s
Host Species Mouse
Immunogen The whole 5.8S ribosomal RNA.
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Test Specificity This is specific for ribosomal RNA. There is no cross-reactivity with other RNAs.
Target Species All species, Mouse, Bacteria, Chicken, Fish, Human, Mouse, Plant, Rat
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG3 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.