Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

rRNA Antibody (Y10b) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 21 publication

$256.00 - $603.56

Specifications

Antigen rRNA
Clone Y10b
Concentration 1.0 mg/mL
Dilution Flow Cytometry 2.5 ug/ml, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34980897), Immunoprecipitation 10 ul/lysate, Immunohistochemistry-Paraffin 1:100 - 1:300, Flow (Intracellular) 2.5 ug/ml, Immunocytochemistry 1 - 3 ug/ml, CyTOF-reported, CyTOF-ready reported by customer review
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100662SS
SDP
View Documents
Novus Biologicals
NB100662SS
0.025 mL
Each for $256.00
Only null left
Add to Cart
 
NB100662
SDP
View Documents
Novus Biologicals
NB100662
0.1 mL
Each for $603.56
Only null left
Add to Cart
 
Description

Description

rRNA Monoclonal specifically detects rRNA in Human, Mouse, All Species, Bacteria samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Intracellular), Immunocytochemistry, CyTOF-reported, CyTOF-ready.
Specifications

Specifications

rRNA
1.0 mg/mL
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
Unconjugated
Mouse
DNA Repair
ribosomal RNA 5.8s
IgG3 κ
Protein A purified
This is specific for ribosomal RNA. There is no cross-reactivity with other RNAs.
Y10b
Flow Cytometry 2.5 ug/ml, Immunohistochemistry 1:100 - 1:300, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 34980897), Immunoprecipitation 10 ul/lysate, Immunohistochemistry-Paraffin 1:100 - 1:300, Flow (Intracellular) 2.5 ug/ml, Immunocytochemistry 1 - 3 ug/ml, CyTOF-reported, CyTOF-ready reported by customer review
Monoclonal
Purified
RUO
Human, Mouse, All species, Bacteria
The whole 5.8S ribosomal RNA.
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.