Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RRP8 Mouse anti-Mouse, Human, Unconjugated, Clone: 5C7.1, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $452.71
Specifications
Antigen | RRP8 |
---|---|
Clone | 5C7.1 |
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABE1013-25
|
MilliporeSigma
MABE101325UG |
25 μg |
Each for $192.35
|
|
|||||
MABE1013-100
|
MilliporeSigma
MABE1013100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-RRP8, clone 5C7.1, Cat. No. MABE1013, is a highly specific mouse monoclonal antibody, that targets Ribosomal RNA-processing protein 8 and has been tested for use in Immunocytochemistry and Western Blotting.Specifications
RRP8 | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Human | |
Ribosomal RNA-processing protein 8;Cerebral protein 1;Nucleomethylin | |
RRP8;KIAA0409;NML;hucep-1 | |
IgG2a κ | |
Protein G purified | |
Clone 5C7.1 specifically detects Ribosomal RNA-processing protein 8. It targets an epitope with in the C-terminal half. | |
∽53 kDa observed; 50.72 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
5C7.1 | |
Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Epigenetics & Nuclear Function | |
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_056139 | |
GST-tagged recombinant fragment corresponding to 98 amino acids from the C-terminal half of human Ribosomal RNA-processing protein 8. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
Antibodies |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title