Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RS 102895 hydrochloride
GREENER_CHOICE

Catalog No. 208910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-891-0 10 mg
20-895-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 20-891-0 Supplier Tocris Bioscience™ Supplier No. 2089/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

CCR2b chemokine receptor antagonist

CCR2-selective chemokine receptor antagonist (IC50 values are 0.36 and 17.8 μM for inhibition of human recombinant CCR2b and CCR1 receptors respectively). Blocks MCP-1-stimulated calcium influx and chemotaxis with IC50 values of 32 nM and 1.7 μM respectively. Also inhibits α1A, α1D and 5-HT1A receptors.

Chemical Identifiers

CAS 1173022-16-6
Molecular Formula C21H22ClF3N2O2

Specifications

Quantity 10 mg
Formula Weight 426.86
Purity >99%
Molecular Weight (g/mol) 426.864
Chemical Name or Material RS 102895 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.