Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RS 127445 hydrochloride
GREENER_CHOICESDP

Catalog No. 299310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-931-0 10 mg
29-935-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-931-0 Supplier Tocris Bioscience™ Supplier No. 2993/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity and selective 5-HT2B antagonist

High affinity 5-HT2B receptor antagonist (pKi = 9.5). Displays 1000-fold selectivity for 5-HT2B with good bioavailability.

Chemical Identifiers

CAS 199864-86-3
Molecular Formula C17H17ClFN3

Specifications

Quantity 10 mg
Formula Weight 317.79
Purity >98%
Molecular Weight (g/mol) 317.79
Chemical Name or Material RS 127445 hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.