Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (RS)-AMPA hydrobromide
GREENER_CHOICESDP

Catalog No. 10741 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-741 1 mg
10-741-0 10 mg
10-745-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 10-741 Supplier Tocris Bioscience™ Supplier No. 1074/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective AMPA agonist; more water soluble form of (RS)-AMPA (Cat. No. 0169)

More water soluble hydrobromide salt of (RS)-AMPA (Cat. No. 0169).

Chemical Identifiers

CAS 171259-81-7
Molecular Formula C7H11BrN2O4

Specifications

Quantity 1 mg
Formula Weight 267.08
Purity >99%
Molecular Weight (g/mol) 267.079
Chemical Name or Material (RS)-AMPA hydrobromide
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.