Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (RS)-AMPA hydrobromide
GREENER_CHOICE

Catalog No. p-4783111
Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective AMPA agonist; more water soluble form of (RS)-AMPA (Cat. No. 0169)

More water soluble hydrobromide salt of (RS)-AMPA (Cat. No. 0169).

Chemical Identifiers

CAS 171259-81-7
Molecular Formula C7H11BrN2O4

Specifications

Quantity 1 mg
Formula Weight 267.08
Purity >99%
Molecular Weight (g/mol) 267.079
Chemical Name or Material (RS)-AMPA hydrobromide
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.