Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ (RS)-MCPG disodium salt

Click to view available options
Quantity:
10 mg
50 mg
Description
(RS)-MCPG disodium salt is a sodium salt of (RS)-MCPG, a non-selective group I/group II metabotropic glutamate receptor antagonist.
S-enantiomer also available.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | (RS)-MCPG disodium salt |
| Purity | >99% |
| Molecular Weight (g/mol) | 253.16 |
| Inhibitors | (RS)-MCPG disodium salt |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction