Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (RS)-MCPG
GREENER_CHOICE

Catalog No. 033650
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-selective mGlu antagonist

Non-selective group I/group II metabotropic glutamate receptor antagonist.

S-enantiomer and Sodium Salt also available.

Chemical Identifiers

CAS 146669-29-6
Molecular Formula C10H11NO4

Specifications

Quantity 50 mg
Formula Weight 209.2
Purity >99%
Molecular Weight (g/mol) 209.201
Chemical Name or Material (RS)-MCPG
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.