Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ (RS)-MCPG

Click to view available options
Quantity:
10 mg
50 mg
Description
Non-selective group I/group II metabotropic glutamate receptor antagonist.
S-enantiomer and Sodium Salt also available.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 209.2 |
| Purity | >99% |
| Molecular Weight (g/mol) | 209.201 |
| Chemical Name or Material | (RS)-MCPG |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction