Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | RS1 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119394
|
Invitrogen™
PA5119394 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121633
|
Invitrogen™
PA5121633 |
20 μL |
Each for $209.50
|
|
|||||
Description
RS1 Polyclonal Antibody for Western Blot, ICC/IF, IHC
May be active in cell adhesion processes during retinal development.Specifications
RS1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human, Pig | |
RS1 | |
retinoschisin; retinoschisin 1; retinoschisis (X-linked, juvenile) 1 (human); retinoschisis 1 homolog; RS; RS1; Rs1h; tmgc1; X-linked juvenile retinoschisis protein; X-linked juvenile retinoschisis protein homolog; XLRS1 | |
RS1 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
O15537 | |
106506914, 6247 | |
Recombinant protein Retinoschisin. The antigen corresponds to amino acid range 24-224 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title